Tisk…

4-Chloro-2-fluoro-5-methylphenylacetonitrile 97%

Dodavatel: Apollo Scientific

PC303323-1G PC303323-5G
APOSPC303323-1GEA 8830 CZK
APOSPC303323-1G APOSPC303323-5G
4-Chloro-2-fluoro-5-methylphenylacetonitrile 97%
4-Chloro-2-fluoro-5-methylphenylacetonitrile
Vzorec: C₉H₇ClFN
MW: 183,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20483139
CAS číslo: 1242314-99-3
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR