Tisk…
3,6-Difluoro-2-methoxybenzyl alcohol 97%
Dodavatel: Apollo Scientific
PC303314-5G PC303314-1G
APOSPC303314-5GEA 20930 CZK
APOSPC303314-5G APOSPC303314-1G
3,6-Difluoro-2-methoxybenzyl alcohol 97%
3,6-Difluoro-2-methoxybenzyl alcohol
Vzorec: C₈H₈F₂O₂
MW: 174.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394897
CAS číslo: 1261746-67-1