2,4-Difluoro-5-methylbenzyl alcohol 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC303302-1G
PC303302-5G
APOSPC303302-1GEA
7940
CZK
APOSPC303302-1G
APOSPC303302-5G
2,4-Difluoro-5-methylbenzyl alcohol 98%
2,4-Difluoro-5-methylbenzyl alcohol
Vzorec:
C₈H₈F₂O MW: 158.145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12196966 CAS číslo: 315204-46-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...