Tisk…

4-Ethyl-2,3-difluorobenzoic acid 97%

Dodavatel: Apollo Scientific

PC303296-5G PC303296-1G
APOSPC303296-5GEA 13460 CZK
APOSPC303296-5G APOSPC303296-1G
4-Ethyl-2,3-difluorobenzoic acid 97%
4-Ethyl-2,3-difluorobenzoic acid
Vzorec: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29277470
CAS číslo: 1429636-88-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR