6-Ethoxy-2,3,4-trifluorobenzyl bromide 97%
Dodavatel: Apollo Scientific
PC303290-1G
PC303290-250MG
APOSPC303290-1GEA
16370
CZK
APOSPC303290-1G
APOSPC303290-250MG
6-Ethoxy-2,3,4-trifluorobenzyl bromide 97%
6-Ethoxy-2,3,4-trifluorobenzyl bromide
Vzorec:
C₉H₈BrF₃O MW: 269,058 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29277488 CAS číslo: 1980054-21-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...