6-Chloro-2,4-difluoro-3-methylbenzyl bromide 97%
Dodavatel: Apollo Scientific
PC303281-1G
PC303281-250MG
APOSPC303281-1GEA
30450
CZK
APOSPC303281-1G
APOSPC303281-250MG
6-Chloro-2,4-difluoro-3-methylbenzyl bromide 97%
6-Chloro-2,4-difluoro-3-methylbenzyl bromide
Vzorec:
C₈H₆BrClF₂ MW: 255.487 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29038429 CAS číslo: 1858251-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...