6-Chloro-2,4-difluoro-3-methylbenzyl alcohol 97%
Dodavatel: Apollo Scientific
PC303280-1G
PC303280-250MG
APOSPC303280-1GEA
27250
CZK
APOSPC303280-1G
APOSPC303280-250MG
6-Chloro-2,4-difluoro-3-methylbenzyl alcohol 97%
6-Chloro-2,4-difluoro-3-methylbenzyl alcohol
Vzorec:
C₈H₇ClF₂O MW: 192,59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29038428 CAS číslo: 1858240-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...