Tisk…

4-Methyl-2,5-bis(trifluoromethyl)benzoic acid 97%

Dodavatel: Apollo Scientific

PC303239-1G PC303239-250MG
APOSPC303239-1GEA 27250 CZK
APOSPC303239-1G APOSPC303239-250MG
4-Methyl-2,5-bis(trifluoromethyl)benzoic acid 97%
4-Methyl-2,5-bis(trifluoromethyl)benzoic acid
Vzorec: C₁₀H₆F₆O₂
MW: 272,144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986831
CAS číslo: 1706436-17-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR