Tisk…

2-Fluoro-4-methyl-3-(trifluoromethyl)phenylacetic acid 97%

Dodavatel: Apollo Scientific

PC303237-1G PC303237-250MG
APOSPC303237-1GEA 27250 CZK
APOSPC303237-1G APOSPC303237-250MG
2-Fluoro-4-methyl-3-(trifluoromethyl)phenylacetic acid 97%
2-Fluoro-4-methyl-3-(trifluoromethyl)phenylacetic acid
Vzorec: C₁₀H₈F₄O₂
MW: 236.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103373
CAS číslo: 1823577-71-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR