Tisk…

2-Methyl-5-(trifluoromethoxy)benzenesulphonyl chloride 97%

Dodavatel: Apollo Scientific

PC303230-1G PC303230-250MG
APOSPC303230-1GEA 25630 CZK
APOSPC303230-1G APOSPC303230-250MG
2-Methyl-5-(trifluoromethoxy)benzenesulphonyl chloride 97%
2-Methyl-5-(trifluoromethoxy)benzenesulphonyl chloride
Vzorec: C₈H₆ClF₃O₃S
Storage Temperature: Ambient
MDL Number: MFCD18399733
CAS číslo: 1261852-46-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR