Tisk…

6-Chloro-2,4-difluoro-3-methylbenzaldehyde 98%

Dodavatel: Apollo Scientific

PC303218-1G PC303218-5G
APOSPC303218-1GEA 5690 CZK
APOSPC303218-1G APOSPC303218-5G
6-Chloro-2,4-difluoro-3-methylbenzaldehyde 98%
6-Chloro-2,4-difluoro-3-methylbenzaldehyde
Vzorec: C₈H₅ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD27986855
CAS číslo: 1706435-05-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR