3-Fluoro-4-methyl-5-(trifluoromethyl)aniline 98%
Dodavatel: Apollo Scientific
PC303207-1G
PC303207-250MG
APOSPC303207-1GEA
22450
CZK
APOSPC303207-1G
APOSPC303207-250MG
3-Fluoro-4-methyl-5-(trifluoromethyl)aniline 98%
3-Fluoro-4-methyl-5-(trifluoromethyl)aniline
Vzorec:
C₈H₇F₄N Storage Temperature: Ambient |
MDL Number:
MFCD27986853 CAS číslo: 1706430-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...