2-Fluoro-4-methyl-3-(trifluoromethyl)benzamide 97%
Dodavatel: Apollo Scientific
PC303192-1G
PC303192-250MG
APOSPC303192-1GEA
20830
CZK
APOSPC303192-1G
APOSPC303192-250MG
2-Fluoro-4-methyl-3-(trifluoromethyl)benzamide 97%
2-Fluoro-4-methyl-3-(trifluoromethyl)benzamide
Vzorec:
C₉H₇F₄NO MW: 221,152 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28103366 CAS číslo: 1824271-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...