Tisk…

2-Chloro-5-methoxy-3-(trifluoromethyl)aniline 97%

Dodavatel: Apollo Scientific

PC303191-1G PC303191-250MG
APOSPC303191-1GEA 19840 CZK
APOSPC303191-1G APOSPC303191-250MG
2-Chloro-5-methoxy-3-(trifluoromethyl)aniline 97%
2-Chloro-5-methoxy-3-(trifluoromethyl)aniline
Vzorec: C₈H₇ClF₃NO
MW: 225,595 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986849
CAS číslo: 1706436-22-7
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR