Tisk…

3-Chloro-4-(trifluoromethyl)benzenesulphonyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC303188-1G PC303188-250MG
APOSPC303188-1GEA 17200 CZK
APOSPC303188-1G APOSPC303188-250MG
3-Chloro-4-(trifluoromethyl)benzenesulphonyl chloride 97%
3-Chloro-4-(trifluoromethyl)benzenesulphonyl chloride
Vzorec: C₇H₃Cl₂F₃O₂S
MW: 279,064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12827565
CAS číslo: 132481-85-7
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR