Tisk…

3,4-Dichloro-5-(trifluoromethoxy)benzamide 97%

Dodavatel: Apollo Scientific

PC303183-1G PC303183-250MG
APOSPC303183-1GEA 19250 CZK
APOSPC303183-1G APOSPC303183-250MG
3,4-Dichloro-5-(trifluoromethoxy)benzamide 97%
3,4-Dichloro-5-(trifluoromethoxy)benzamide
Vzorec: C₈H₄Cl₂F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD28054196
CAS číslo: 1706435-10-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR