Tisk…

3,4-Dichloro-5-(trifluoromethoxy)benzyl alcohol 97%

Dodavatel: Apollo Scientific

PC303173-1G PC303173-250MG
APOSPC303173-1GEA 14490 CZK
APOSPC303173-1G APOSPC303173-250MG
3,4-Dichloro-5-(trifluoromethoxy)benzyl alcohol 97%
3,4-Dichloro-5-(trifluoromethoxy)benzyl alcohol
Vzorec: C₈H₅Cl₂F₃O₂
MW: 261.025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28054206
CAS číslo: 1706458-54-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR