3,4-Dichloro-5-(trifluoromethyl)benzoyl chloride 97%
Dodavatel: Apollo Scientific
PC303167-1G
PC303167-250MG
APOSPC303167-1GEA
16670
CZK
APOSPC303167-1G
APOSPC303167-250MG
3,4-Dichloro-5-(trifluoromethyl)benzoyl chloride 97%
3,4-Dichloro-5-(trifluoromethyl)benzoyl chloride
Vzorec:
C₈H₂Cl₃F₃O MW: 277,455 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28054199 CAS číslo: 1706458-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...