2-Fluoro-4-methyl-3-(trifluoromethyl)phenol 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC303165-1G
PC303165-250MG
APOSPC303165-1GEA
16670
CZK
APOSPC303165-1G
APOSPC303165-250MG
2-Fluoro-4-methyl-3-(trifluoromethyl)phenol 98%
2-Fluoro-4-methyl-3-(trifluoromethyl)phenol
Vzorec:
C₈H₆F₄O Storage Temperature: Ambient |
MDL Number:
MFCD28103361 CAS číslo: 1823323-63-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...