4-Fluoro-3-methyl-2-(trifluoromethyl)benzyl alcohol 98%
Dodavatel: Apollo Scientific
PC303161-1G
PC303161-250MG
APOSPC303161-1GEA
15380
CZK
APOSPC303161-1G
APOSPC303161-250MG
4-Fluoro-3-methyl-2-(trifluoromethyl)benzyl alcohol 98%
4-Fluoro-3-methyl-2-(trifluoromethyl)benzyl alcohol
Vzorec:
C₉H₈F₄O MW: 208,153 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28054201 CAS číslo: 1706458-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...