Tisk…

3,4-Dichloro-5-(trifluoromethoxy)benzaldehyde 97%

Dodavatel: Apollo Scientific
Varování

PC303158-1G PC303158-250MG
APOSPC303158-1GEA 15380 CZK
APOSPC303158-1G APOSPC303158-250MG
3,4-Dichloro-5-(trifluoromethoxy)benzaldehyde 97%
3,4-Dichloro-5-(trifluoromethoxy)benzaldehyde
Vzorec: C₈H₃Cl₂F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD28054205
CAS číslo: 1706430-32-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR