Tisk…

3,4-Dichloro-5-(trifluoromethoxy)benzoic acid 97%

Dodavatel: Apollo Scientific

PC303157-1G PC303157-250MG
APOSPC303157-1GEA 14880 CZK
APOSPC303157-1G APOSPC303157-250MG
3,4-Dichloro-5-(trifluoromethoxy)benzoic acid 97%
3,4-Dichloro-5-(trifluoromethoxy)benzoic acid
Vzorec: C₈H₃Cl₂F₃O₃
MW: 275,009 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28054192
CAS číslo: 1706458-45-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR