2-Fluoro-4-methyl-3-(trifluoromethyl)benzaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC303154-1G
PC303154-250MG
APOSPC303154-1GEA
14450
CZK
APOSPC303154-1G
APOSPC303154-250MG
2-Fluoro-4-methyl-3-(trifluoromethyl)benzaldehyde 97%
2-Fluoro-4-methyl-3-(trifluoromethyl)benzaldehyde
Vzorec:
C₉H₆F₄O MW: 206,137 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28103358 CAS číslo: 1824270-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...