3,4-Dichloro-5-(trifluoromethyl)benzoic acid 97%
Dodavatel: Apollo Scientific
PC303153-1G
PC303153-250MG
APOSPC303153-1GEA
14060
CZK
APOSPC303153-1G
APOSPC303153-250MG
3,4-Dichloro-5-(trifluoromethyl)benzoic acid 97%
3,4-Dichloro-5-(trifluoromethyl)benzoic acid
Vzorec:
C₈H₃Cl₂F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD28054189 CAS číslo: 1706458-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...