Tisk…

3-Butoxy-2,6-difluorobenzyl alcohol 98%

Dodavatel: Apollo Scientific

PC303141-10G PC303141-5G
APOSPC303141-10GEA 16040 CZK
APOSPC303141-10G APOSPC303141-5G
3-Butoxy-2,6-difluorobenzyl alcohol 98%
3-Butoxy-2,6-difluorobenzyl alcohol
Vzorec: C₁₁H₁₄F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD27986841
CAS číslo: 1706435-02-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR