Tisk…

4-Methoxy-3-methyl-5-(trifluoromethyl)phenylacetic acid 97%

Dodavatel: Apollo Scientific

PC303138-1G PC303138-250MG
APOSPC303138-1GEA 20240 CZK
APOSPC303138-1G APOSPC303138-250MG
4-Methoxy-3-methyl-5-(trifluoromethyl)phenylacetic acid 97%
4-Methoxy-3-methyl-5-(trifluoromethyl)phenylacetic acid
Vzorec: C₁₁H₁₁F₃O₃
MW: 248,198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986835
CAS číslo: 1706446-17-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR