Tisk…

3-Fluoro-2-methoxyphenylacetonitrile 98%

Dodavatel: Apollo Scientific

PC303124-1G PC303124-5G
APOSPC303124-1GEA 7080 CZK
APOSPC303124-1G APOSPC303124-5G
3-Fluoro-2-methoxyphenylacetonitrile 98%
3-Fluoro-2-methoxyphenylacetonitrile
Vzorec: C₉H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD09832388
CAS číslo: 1017779-73-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR