Tisk…

4-Chloro-3,5-difluorobenzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

PC303081-10G PC303081-5G PC303081-1G
APOSPC303081-10GEA 18320 CZK
APOSPC303081-10G APOSPC303081-5G APOSPC303081-1G
4-Chloro-3,5-difluorobenzyl bromide
4-Chloro-3,5-difluorobenzyl bromide
Vzorec: C₇H₄BrClF₂
MW: 241,46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23795325
CAS číslo: 1400991-56-1
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR