3-Chloro-5-(trifluoromethoxy)benzamide 98%
Dodavatel: Apollo Scientific
PC303059-1G
PC303059-250MG
APOSPC303059-1GEA
0
CZK
APOSPC303059-1G
APOSPC303059-250MG
3-Chloro-5-(trifluoromethoxy)benzamide 98%
3-Chloro-5-(trifluoromethoxy)benzamide
Vzorec:
C₈H₅ClF₃NO₂ MW: 239,579 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660308 CAS číslo: 886503-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...