2-Ethoxy-3,5-difluorobenzyl bromide 97%
Dodavatel: Apollo Scientific
PC303032-10G
PC303032-5G
PC303032-1G
APOSPC303032-10GEA
12700
CZK
APOSPC303032-10G
APOSPC303032-5G
APOSPC303032-1G
2-Ethoxy-3,5-difluorobenzyl bromide 97%
2-Ethoxy-3,5-difluorobenzyl bromide
Vzorec:
C₉H₉BrF₂O MW: 251.068 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09258718 CAS číslo: 1017779-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...