Tisk…

2,6-Dichloro-4-(pentafluorosulphur)nitrobenzene 97%

Dodavatel: Apollo Scientific

PC303021-1G
APOSPC303021-1GEA 35380 CZK
APOSPC303021-1G
2,6-Dichloro-4-(pentafluorosulphur)nitrobenzene 97%
2,6-Dichloro-4-(pentafluorosulphur)nitrobenzene
Vzorec: C₆H₂Cl₂F₅NO₂S
MW: 318,049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23795321
CAS číslo: 1431329-68-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR