Tisk…

4-Fluoro-3-methylphenylacetonitrile 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (4-Fluoro-3-methylphenyl)acetonitrile

PC302996-10G PC302996-5G PC302996-1G
APOSPC302996-10GEA 10250 CZK
APOSPC302996-10G APOSPC302996-5G APOSPC302996-1G
4-Fluoro-3-methylphenylacetonitrile 98%
4-Fluoro-3-methylphenylacetonitrile
Vzorec: C₉H₈FN
Storage Temperature: Ambient
MDL Number: MFCD09832240
CAS číslo: 1000548-41-3
UN: 3276
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR