4'-Chloro-3',5'-difluoroacetophenone 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC302980-5G
PC302980-1G
APOSPC302980-5GEA
16370
CZK
APOSPC302980-5G
APOSPC302980-1G
4'-Chloro-3',5'-difluoroacetophenone 98%
4'-Chloro-3',5'-difluoroacetophenone
Vzorec:
C₈H₅ClF₂O MW: 190,575 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11553418 CAS číslo: 1256352-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...