Tisk…

4-Chloro-3-(trifluoromethoxy)benzyl alcohol 97%

Dodavatel: Apollo Scientific

PC302978-10G PC302978-5G PC302978-25G
APOSPC302978-10GEA 5390 CZK
APOSPC302978-10G APOSPC302978-5G APOSPC302978-25G
4-Chloro-3-(trifluoromethoxy)benzyl alcohol 97%
4-Chloro-3-(trifluoromethoxy)benzyl alcohol
Vzorec: C₈H₆ClF₃O₂
MW: 226,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660248
CAS číslo: 886500-89-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR