3-Ethoxy-2,6-difluorobenzyl bromide 98%
Dodavatel: Apollo Scientific
PC302953-10G
PC302953-5G
PC302953-1G
APOSPC302953-10GEA
9000
CZK
APOSPC302953-10G
APOSPC302953-5G
APOSPC302953-1G
3-Ethoxy-2,6-difluorobenzyl bromide 98%
3-Ethoxy-2,6-difluorobenzyl bromide
Vzorec:
C₉H₉BrF₂O MW: 251,068 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11519372 CAS číslo: 1092461-30-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...