Tisk…

3-Chloro-5-fluoro-4-methoxycinnamic acid

Dodavatel: Apollo Scientific

PC302951-10G PC302951-5G
APOSPC302951-10GEA 11540 CZK
APOSPC302951-10G APOSPC302951-5G
3-Chloro-5-fluoro-4-methoxycinnamic acid
3-Chloro-5-fluoro-4-methoxycinnamic acid
Vzorec: C₁₀H₈ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD04116009
CAS číslo: 886501-39-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR