3-(3,4,5-Trifluorophenyl)propionic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(3,4,5-Trifluorophenyl)propanoic acid
PC302938-10G
PC302938-5G
PC302938-1G
APOSPC302938-10GEA
9000
CZK
APOSPC302938-10G
APOSPC302938-5G
APOSPC302938-1G
3-(3,4,5-Trifluorophenyl)propionic acid 97%
3-(3,4,5-Trifluorophenyl)propionic acid
Vzorec:
C₉H₇F₃O₂ MW: 204,146 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660216 CAS číslo: 886499-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...