Tisk…

3-(2,4,5-Trifluorophenyl)propionic acid 98%

Dodavatel: Apollo Scientific

PC302937-10G PC302937-5G PC302937-1G
APOSPC302937-10GEA 8770 CZK
APOSPC302937-10G APOSPC302937-5G APOSPC302937-1G
3-(2,4,5-Trifluorophenyl)propionic acid 98%
3-(2,4,5-Trifluorophenyl)propionic acid
Vzorec: C₉H₇F₃O₂
MW: 204,146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660217
CAS číslo: 651047-33-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR