Tisk…

2-Methoxy-5-(trifluoromethoxy)phenylacetonitrile 97%

Dodavatel: Apollo Scientific

PC302928-10G PC302928-5G PC302928-1G
APOSPC302928-10GEA 13160 CZK
APOSPC302928-10G APOSPC302928-5G APOSPC302928-1G
2-Methoxy-5-(trifluoromethoxy)phenylacetonitrile 97%
2-Methoxy-5-(trifluoromethoxy)phenylacetonitrile
Vzorec: C₁₀H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD04972914
CAS číslo: 886503-57-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR