Tisk…

6-Ethoxy-2,3-difluorobenzaldehyde 98%

Dodavatel: Apollo Scientific

PC302900-10G PC302900-5G PC302900-1G
APOSPC302900-10GEA 7710 CZK
APOSPC302900-10G APOSPC302900-5G APOSPC302900-1G
6-Ethoxy-2,3-difluorobenzaldehyde 98%
6-Ethoxy-2,3-difluorobenzaldehyde
Vzorec: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD13181644
CAS číslo: 167684-02-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR