Tisk…

5-Fluoro-2-(trifluoromethoxy)phenylacetonitrile 97%

Dodavatel: Apollo Scientific
Varování

PC302891-1G PC302891-5G PC302891-10G
APOSPC302891-1GEA 4530 CZK
APOSPC302891-1G APOSPC302891-5G APOSPC302891-10G
5-Fluoro-2-(trifluoromethoxy)phenylacetonitrile 97%
5-Fluoro-2-(trifluoromethoxy)phenylacetonitrile
Vzorec: C₉H₅F₄NO
MW: 219,136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519338
CAS číslo: 1092460-90-6
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR