3-Chloro-5-fluoro-4-methoxybenzyl alcohol 97%
Dodavatel: Apollo Scientific
PC302773-10G
PC302773-5G
APOSPC302773-10GEA
9990
CZK
APOSPC302773-10G
APOSPC302773-5G
3-Chloro-5-fluoro-4-methoxybenzyl alcohol 97%
3-Chloro-5-fluoro-4-methoxybenzyl alcohol
Vzorec:
C₈H₈ClFO₂ MW: 190.599 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115867 CAS číslo: 886497-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...