5-Chloro-2-(trifluoromethoxy)benzylamine 98%
Dodavatel: Apollo Scientific
PC302759-1G
PC302759-5G
PC302759-250MG
APOSPC302759-1GEA
11380
CZK
APOSPC302759-1G
APOSPC302759-5G
APOSPC302759-250MG
5-Chloro-2-(trifluoromethoxy)benzylamine 98%
5-Chloro-2-(trifluoromethoxy)benzylamine
Vzorec:
C₈H₇ClF₃NO Storage Temperature: Ambient |
MDL Number:
MFCD11519365 CAS číslo: 874821-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...