Tisk…

3-Chloro-5-fluorocinnamic acid 95%

Dodavatel: Apollo Scientific

PC302719-5G PC302719-1G
APOSPC302719-5GEA 3870 CZK
APOSPC302719-5G APOSPC302719-1G
3-Chloro-5-fluorocinnamic acid 95%
3-Chloro-5-fluorocinnamic acid
Vzorec: C₉H₆ClFO₂
MW: 200,594 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116028
CAS číslo: 936366-61-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR