Tisk…

3-[4-Methyl-3-(trifluoromethyl)phenyl]propionic acid 98%

Dodavatel: Apollo Scientific

PC302707-1G PC302707-5G PC302707-10G
APOSPC302707-1GEA 5790 CZK
APOSPC302707-1G APOSPC302707-5G APOSPC302707-10G
3-[4-Methyl-3-(trifluoromethyl)phenyl]propionic acid 98%
3-[4-Methyl-3-(trifluoromethyl)phenyl]propionic acid
Vzorec: C₁₁H₁₁F₃O₂
MW: 232,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832295
CAS číslo: 1017778-20-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR