3-[2-Methyl-3-(trifluoromethyl)phenyl]propionic acid 98%
Dodavatel: Apollo Scientific
PC302705-1G
PC302705-5G
PC302705-10G
APOSPC302705-1GEA
4240
CZK
APOSPC302705-1G
APOSPC302705-5G
APOSPC302705-10G
3-[2-Methyl-3-(trifluoromethyl)phenyl]propionic acid 98%
3-[2-Methyl-3-(trifluoromethyl)phenyl]propionic acid
Vzorec:
C₁₁H₁₁F₃O₂ MW: 232,199 g/mol Bod tání: 90…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD09832282 CAS číslo: 1017777-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...