Tisk…

3-Bromo-4-fluorophenylsulphur pentafluoride 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-5-(pentafluorosulphur)bromobenzene

PC302686-1G PC302686-5G
APOSPC302686-1GEA 10910 CZK
APOSPC302686-1G APOSPC302686-5G
3-Bromo-4-fluorophenylsulphur pentafluoride 97%
3-Bromo-4-fluorophenylsulphur pentafluoride
Vzorec: C₆H₃BrF₆S
MW: 301,047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986829
CAS číslo: 1706434-97-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR