Tisk…

2,6-Difluoro-4-methoxybenzyl alcohol 98%

Dodavatel: Apollo Scientific

PC302671-10G PC302671-5G
APOSPC302671-10GEA 8870 CZK
APOSPC302671-10G APOSPC302671-5G
2,6-Difluoro-4-methoxybenzyl alcohol 98%
2,6-Difluoro-4-methoxybenzyl alcohol
Vzorec: C₈H₈F₂O₂
MW: 174.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115923
CAS číslo: 79538-27-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR