2',6'-Difluoro-3'-methoxyacetophenone 98%
Dodavatel: Apollo Scientific
Synonyms:
1-(2,6-Difluoro-3-methoxyphenyl)ethanone
PC302666-10G
PC302666-5G
PC302666-1G
APOSPC302666-10GEA
12900
CZK
APOSPC302666-10G
APOSPC302666-5G
APOSPC302666-1G
2',6'-Difluoro-3'-methoxyacetophenone 98%
2',6'-Difluoro-3'-methoxyacetophenone
Vzorec:
C₉H₈F₂O₂ MW: 186,155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20487626 CAS číslo: 1206593-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...