Tisk…

2,4-Difluoro-3-methoxybenzyl alcohol 97%

Dodavatel: Apollo Scientific

Synonyms: (2,4-difluoro-3-methoxyphenyl)methanol

PC302663-5G PC302663-1G
APOSPC302663-5GEA 6450 CZK
APOSPC302663-5G APOSPC302663-1G
2,4-Difluoro-3-methoxybenzyl alcohol 97%
2,4-Difluoro-3-methoxybenzyl alcohol
Vzorec: C₈H₈F₂O₂
MW: 174,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115934
CAS číslo: 178974-98-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR